missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIA/ALK-2/Activin Receptor Type 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-90255
This item is not returnable.
View return policy
Description
Activin RIA/ALK-2/Activin Receptor Type 1 Polyclonal antibody specifically detects Activin RIA/ALK-2/Activin Receptor Type 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Activin RIA/ALK-2/Activin Receptor Type 1 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| activin A receptor, type I, ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RRNQERLNPRDVEYGTIEGLITTNVGDSTLADLLDHSCTSGSGSGLPFLVQRTVARQITLLE | |
| 0.1 mL | |
| Protein Kinase | |
| 90 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction