Learn More
TMEM169 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68790
 View more versions of this product
Product Code. 18691158
  • ONLINE EXCLUSIVE PRICE
    € 624.00
    € 590.10 / 100µL
    Save € 33.90
    5% Off
Description
TMEM169 Polyclonal antibody specifically detects TMEM169 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEM169
Polyclonal
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
DKFZp781L2456, FLJ34263, transmembrane protein 169
This antibody was developed against a recombinant protein corresponding to amino acids: VVQLSWSWEAWWQAARDMEKGFCGWLCSKLGLEDCSPYSIVELLESDNISSTLSNKDPIQEVETST
100 μg
Primary
Human
Purified
Immunofluorescence
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Protein A purified
RUO
92691
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.