Learn More
Invitrogen™ TMEM16K Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139762
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ATLLITSQIL NQIMESFLPY WLQRKHGVRV KRKVQALKAD IDATLYEQVI LEKEMGTYLG TFDDYLEL Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 84%.
TMEM16K is a multi-pass membrane protein. It belongs to the anoctamin family. TMEM16K may act as a calcium-activated chloride channel.
Specifications
| TMEM16K | |
| Polyclonal | |
| Unconjugated | |
| Ano10 | |
| AI604832; ANO10; ano10a; anoctamin 10; anoctamin 10a; anoctamin-10; fc37b04; hypothetical protein LOC566425; RGD1308260; SCAR10; Tmem16k; Transmembrane protein 16K; wu:fc37b04; zgc:114140 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NW15 | |
| Ano10 | |
| Recombinant protein corresponding to Human ANO10. Recombinant protein control fragment (Product #RP-91168). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.