missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM173 Monoclonal Antibody (OTI4H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505024S
This item is not returnable.
View return policy
Description
TMEM173 Monoclonal antibody specifically detects TMEM173 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TMEM173 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86WV6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 340061 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H8 | |
| Unconjugated | |
| TMEM173 | |
| 2610307O08Rik; endoplasmic reticulum IFN stimulator; endoplasmic reticulum interferon stimulator; ERIS; hMITA; hSTING; hypothetical LOC533661; mediator of IRF3 activation; Mita; mitochondrial mediator of IRF3 activation; mitochondrial transmembrane protein 173; MMITA; Mpys; mSTING; NET23; N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner; poSTING; RGD1562552; rSTING; SAVI; Stimulator of interferon genes protein; stimulator of interferon response cGAMP interactor 1; STING; STING1; tm173; Tmem173; Transmembrane protein 173 | |
| Full length human recombinant protein of human TMEM173 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction