Learn More
TMEM173 Mouse anti-Human, Clone: OTI1G5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505018
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.Specifications
| TMEM173 | |
| Monoclonal | |
| Unconjugated | |
| TMEM173 | |
| ERIS, MITA, MPYS, NET23, SAVI, STING, hMITA, hSTING | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 340061 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI1G5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q86WV6 | |
| TMEM173 | |
| Full length human recombit protein of human TMEM173 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.