missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM181 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55411-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to TMEM181.
Mediates action of cytolethal distending toxins (CDT), which are secreted by many pathogenic bacteria. Expression level of TMEM181 is rate-limiting for intoxication.Specifications
| TMEM181 | |
| Polyclonal | |
| Unconjugated | |
| TMEM181 | |
| GPR178, KIAA1423, TMEM181, transmembrane protein 181 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57583, 77106 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9P2C4 | |
| TMEM181, Tmem181a | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction