missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM184A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596834
This item is not returnable.
View return policy
Description
Immunogen sequence: FPCQVYAEKK ENSPAPPAPM QSISSGIRET VSPQDIVQDA IHNFSPAYQH YTQQATHEAP RPGTHPSGGS GGSRKSRSLE KRMLIPSEDL; Positive Samples: Mouse lung, Mouse liver, Mouse kidney, Rat lung; Cellular Location: Membrane, Multi-pass membrane protein.
TMEM184A, a multi-pass transmembrane protein, belongs to the UPF0206 family. While the exact function of TMEM184A remains unclear, it is known to be expressed at high levels in the developing testis. TMEM184A may mediate sex-specific signaling events during Sertoli cell differentiation. The restricted expression of this gene in Sertoli cells suggests a role for the encoded protein in the development and/or continued function of that cell type. Ectopic expression of TMEM184A in female genital ridges resulted in failure of gonocytes to enter meiosis.
Specifications
| TMEM184A | |
| Polyclonal | |
| Unconjugated | |
| TMEM184A | |
| RGD1306702; Sdmg1; Sexually dimorphic expressed in male gonads 1; tcag7.540; TMEM184A; Transmembrane protein 184A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 231832, 304325 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UFJ6, Q4QQS1 | |
| TMEM184A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 324-413 of human TMEM184A (NP_0010910891). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction