missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: KLCDHQLARW RLFQCLTGHF WSKVCD Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 35%.
This gene consists of two exons and encodes a multi-pass membrane protein. An alternatively spliced transcript variant encoding the same protein has been found, but its biological validity is not determined.
Specifications
Specifications
| Antigen | TMEM187 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM187 |
| Gene Accession No. | Q14656 |
| Gene Alias | CXorf12; DXS9878E; ITBA1; Protein ITBA1; TMEM187; Transmembrane protein 187 |
| Gene Symbols | TMEM187 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
