missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM192 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-82029CL1
This item is not returnable.
View return policy
Description
TMEM192 Polyclonal antibody specifically detects TMEM192 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| TMEM192 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 201931 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| FLJ38482, transmembrane protein 192 | |
| TMEM192 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human TMEM192. The immunogen is located within amino acids 180 - 230 of TMEM192. Amino Acid Squence: IRRFNKAKPEPDILEEEK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction