Learn More
TMEM192 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98548MFV500
Product Code. 18596400
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TMEM192 Polyclonal antibody specifically detects TMEM192 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mFluor Violet 500 SE
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
FLJ38482, transmembrane protein 192
Store at 4C in the dark.