missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM192 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553221
This item is not returnable.
View return policy
Description
Immunogen sequence: EPDILEEEKI YAYPSNITSE TGFRTISSLE EIVEKQGDTI EYLKRHNALL SKRLLALTSS DLGCQ Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.
Specifications
| TMEM192 | |
| Polyclonal | |
| Unconjugated | |
| TMEM192 | |
| 3110005G23Rik; FLJ38482; RGD1309341; TMEM192; Transmembrane protein 192 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY95 | |
| TMEM192 | |
| Recombinant protein corresponding to Human TMEM192. Recombinant protein control fragment (Product #RP-91176). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction