missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM209 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544926
This item is not returnable.
View return policy
Description
Peptide sequence: MMQGEAHPSA SLIDRTIKMR KETEARKVVL AWGLLNVSMA GMIYTEMTGK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 100%.
TMEM209 (Transmembrane Protein 209) is a Protein Coding gene. Diseases associated with TMEM209 include Hyperalphalipoproteinemia 1. [GeneCards].
Specifications
| TMEM209 | |
| Polyclonal | |
| Unconjugated | |
| TMEM209 | |
| 2700094F01Rik; AI428435; NET31; RGD1309682; testicular tissue protein Li 202; Tmem209; Transmembrane protein 209; Transmembrane protein 209-like protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84928 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96SK2 | |
| TMEM209 | |
| Synthetic peptide directed towards the N-terminal of human FLJ14803 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction