missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a/CD18 Antibody (hu1124 (Efalizumab)), PerCP - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52686PCP
This item is not returnable.
View return policy
Description
CD11a/CD18 Monoclonal antibody specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF, Neutralization
Specifications
| CD11a/CD18 | |
| Monoclonal | |
| PerCP | |
| CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1, alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
| Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 3683 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry, CyTOF, Neutralization | |
| hu1124 (Efalizumab) | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction