missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM248 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06060-100ul
This item is not returnable.
View return policy
Description
TMEM248 Polyclonal antibody specifically detects TMEM248 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TMEM248 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| C7orf42, transmembrane protein 248 | |
| Produced in rabbits immunized with E. coli-derived Human TMEM248 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55069 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion