missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM25 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00184CL1
This item is not returnable.
View return policy
Description
TMEM25 Polyclonal antibody specifically detects TMEM25 in Human samples. It is validated for ELISA
Specifications
| TMEM25 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| CoraFluor 1 | |
| 0610039J01Rik, FLJ14399, transmembrane protein 25 | |
| Produced in rabbits immunized with purified, recombinant Human TMEM25 (Accession#: AAH51841.1; Met1-Pro224) | |
| 0.1 mL | |
| Cell Biology | |
| 84866 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction