missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24361-1-AP-150UL
This item is not returnable.
View return policy
Description
TMEM25 (Transmembrane Protein 25) is a Protein Coding gene. An important paralog of this gene is SIGLEC1.Specifications
| TMEM25 | |
| Polyclonal | |
| Unconjugated | |
| TMEM25 | |
| TMEM25, transmembrane protein 25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 689172, 71687, 84866 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86YD3, Q9DCF1 | |
| TMEM25 | |
| TMEM25 Fusion Protein Ag19964 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction