missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM255A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561745
This item is not returnable.
View return policy
Description
Immunogen sequence: FKDMNPTLPA LNCSVENTHP TVSYYAHPQV ASYNTYYHSP PHLPPYSAYD FQHSGVFPSS PPSGLSDEPQ SASPSPSYMW SSSAPPRYSP PY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Specifications
| TMEM255A | |
| Polyclonal | |
| Unconjugated | |
| TMEM255A | |
| 4933417N17; 6430550H21Rik; FAM70A; family with sequence similarity 70, member A; protein FAM70A; TMEM255A; Transmembrane protein 255A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245386, 313453, 55026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5JRV8, Q7TMP6, Q8BHW5 | |
| TMEM255A | |
| Recombinant protein corresponding to Human TMEM255A. Recombinant protein control fragment (Product #RP-95441). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction