missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562842
This item is not returnable.
View return policy
Description
Immunogen sequence: LMVSQLCEDK HSQCKWVMGS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.
Thought to stabilize the calcium channel in an inactivated (closed) state. Modulates calcium current when coexpressed with CACNA1G.
Specifications
| TMEM37 | |
| Polyclonal | |
| Unconjugated | |
| TMEM37 | |
| AI173373; AW540203; Cacng5; Neuronal voltage-gated calcium channel gamma-like subunit; Pr; Pr1; protein distantly related to to the gamma subunit family; TMEM37; tmem37 {ECO:0000250; Transmembrane protein 37; UniProtKB:Q9JJV3}; voltage-dependent calcium channel gamma subunit-like protein; voltage-dependent calcium channel gamma-like subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WXS4 | |
| TMEM37 | |
| Recombinant protein corresponding to Human TMEM37. Recombinant protein control fragment (Product #RP-99899). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction