missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM38A Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97578JF549
This item is not returnable.
View return policy
Description
TMEM38A Polyclonal specifically detects TMEM38A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TMEM38A | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 79041 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| transmembrane protein 38ATRIC-AMGC3169, TRICA, trimeric intracellular cation channel type A | |
| Produced in rabbits immunized with E. coli-derived Human TMEM38A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction