missing translation for 'onlineSavingsMsg'
Learn More

TMEM43 Polyclonal Antibody
GREENER_CHOICE

TMEM43 Polyclonal Antibody

Brand:  Invitrogen PA576407

Product Code. 15912935

  • € 582.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TMEM43 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMEM43
Polyclonal
Unconjugated
TMEM43
1200015A22Rik; ARVC5; ARVD5; EDMD7; LUMA; Protein LUMA; Tmem43; Transmembrane protein 43; UNQ2564/PRO6244
Rabbit
Antigen affinity chromatography
RUO
74122, 79188
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q9BTV4, Q9DBS1
TMEM43
Recombinant protein of human TMEM43.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.