missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM44 Polyclonal Antibody

Product Code. 15788311 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15788311 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15788311 Supplier Invitrogen™ Supplier No. PA560482

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DTQALLTCAE KEEENQENLD WVPLTTLSHC KSLRTMTAIS RYMELTIEPV QQAGCSATRL PGDGQTSAGD ASLQDPPSYP PVQVIRA Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 76%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM44
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM44
Gene Accession No. Q2T9K0
Gene Alias 1700007N03Rik; 9330161C17Rik; B230220N21; RGD1563737; TMEM44; Transmembrane protein 44
Gene Symbols TMEM44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM44. Recombinant protein control fragment (Product #RP-100528).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93109
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.