missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM54 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | TMEM54 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18206194
|
Novus Biologicals
NBP2-56844 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656336
|
Novus Biologicals
NBP2-56844-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM54 Polyclonal specifically detects TMEM54 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| TMEM54 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 113452 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IAMTFATQGKALLAACTFGSSELLALAPDCPFD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| BCLP, Beta-casein-like protein, CAC1, CAC-1, MGC10137, Protein CAC-1, transmembrane protein 54 | |
| TMEM54 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title