missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524873
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology.
Plays a role in the regulation of neuronal activity.
Specifications
| TMEM57 | |
| Polyclonal | |
| Unconjugated | |
| TMEM57 | |
| 1110007C24Rik; 9230118A01Rik; AI317300; AI606104; brain-specific adapter protein C61; brain-specific adaptor protein C61; C61; MACO1; macoi; Macoilin; Macoilin-1; Tmem57; Transmembrane protein 57 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8N5G2 | |
| TMEM57 | |
| KLH conjugated synthetic peptide between 233-262 amino acids from the N-terminal region of human MACOI. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction