missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM59 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76497AF405
This item is not returnable.
View return policy
Description
TMEM59 Polyclonal antibody specifically detects TMEM59 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TMEM59 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| C1orf8, chromosome 1 open reading frame 8, FLJ42977, HSPC001, Liver membrane-bound protein, transmembrane protein 59 | |
| Antibody was raised against a 16 amino acid synthetic peptide near amino terminus of human TMEM59. The immunogen is located within amino acids 80 - 130 of TMEM59. Amino Acid Squence: ECESACTEAYSQSDEQ | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
| 9528 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction