missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM67 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141155
This item is not returnable.
View return policy
Description
TMEM67 Polyclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene localizes to the primary cilium and to the plasma membrane. The gene functions in centriole migration to the apical membrane and formation of the primary cilium. Multiple transcript variants encoding different isoforms have been found for this gene. Defects in this gene are a cause of Meckel syndrome type 3 (MKS3) and Joubert syndrome type 6 (JBTS6).
Specifications
| TMEM67 | |
| Polyclonal | |
| Unconjugated | |
| TMEM67 | |
| 5330408M12Rik; B230117O07; b2b1163.1Clo; b2b1291.1Clo; JBTS6; meckel syndrome type 3 protein; Meckel syndrome type 3 protein homolog; Meckelin; Mks3; NPHP11; TMEM67; TNEM67; transmembrane protein 67 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 329795, 91147 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5HYA8, Q8BR76 | |
| TMEM67 | |
| Synthetic peptide within human Meckelin aa 20-80. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction