missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM70 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77131UV
This item is not returnable.
View return policy
Description
TMEM70 Polyclonal antibody specifically detects TMEM70 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TMEM70 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| FLJ20533, transmembrane protein 70, transmembrane protein 70, mitochondrial | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human TMEM70. The immunogen is located within amino acids 150 - 200 of TMEM70. Amino Acid Squence: RLYHEATTDTYKAI | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 54968 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction