missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM80 Antibody (OTI3A6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74546AF488
This item is not returnable.
View return policy
Description
TMEM80 Monoclonal specifically detects TMEM80 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| TMEM80 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TMEM80 | |
| Full length recombinant protein of human TMEM80 (NP_777600) produced in HEK293T cells. | |
| 0.1 mL | |
| 283232.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI3A6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| FLJ38216, FLJ55823, transmembrane protein 80 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction