missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM86B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563469
This item is not returnable.
View return policy
Description
Immunogen sequence: MDAGKAGQTL KTHCSAQRPD VC Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 45%.
Specifications
| TMEM86B | |
| Polyclonal | |
| Unconjugated | |
| TMEM86B | |
| alkenylglycerophosphocholine hydrolase; alkenylglycerophosphoethanolamine hydrolase; lysoplasmalogenase; TMEM86B; Transmembrane protein 86B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 255043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N661 | |
| TMEM86B | |
| Recombinant protein corresponding to Human TMEM86B. Recombinant protein control fragment (Product #RP-99209). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction