missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM87A Antibody (772807) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7966JF525
This item is not returnable.
View return policy
Description
TMEM87A Monoclonal antibody specifically detects TMEM87A in Human samples. It is validated for Flow Cytometry,Western Blot,Simple Western,CyTOF
Specifications
| TMEM87A | |
| Monoclonal | |
| Janelia Fluor 525 | |
| DKFZp564G2022, PSEC0094, transmembrane protein 87A | |
| E. coli-derived recombinant human TMEM87A, Ser22-Asn157, Accession # Q8NBN3 | |
| RUO | |
| 25963 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, Simple Western, CyTOF | |
| 772807 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction