missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM97 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92522
This item is not returnable.
View return policy
Description
TMEM97 Polyclonal antibody specifically detects TMEM97 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM97 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| MAC30Protein MAC30, transmembrane protein 97 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FEDFSKASGFKGQRPETLHERLT | |
| 0.1 mL | |
| Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers | |
| 27346 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction