missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM9B Recombinant Rabbit Monoclonal Antibody (K01_2J39)
GREENER_CHOICE

Product Code. 30828647
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828647 100 μL
30830266 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828647 Supplier Invitrogen™ Supplier No. MA563878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TMEM9B Recombinant Monoclonal Antibody for Western Blot

TMEM9B, also termed as Transmembrane protein 9B encodes a 198 amino-acid protein that contains an N-terminal signal peptide, a single transmembrane region, three potential N-glycosylation sites, and three conserved cys-rich domains in the N-terminus. There is a dimer form of TMEM9B protein. TMEM9B mRNA is expressed in a wide range of tissues and cells. TMEM9B is a lysosomal transmembrane protein that regulates the activity of inflammatory signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM9B
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2J39
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene TMEM9B
Gene Accession No. Q9JJR8, Q9NQ34
Gene Alias 2310004K06Rik; AW539847; C11orf15; D7H11orf15; ICRFP703B1614Q5.3; ICRFP703N2430Q5.3; RGD1310775; TMEM9 domain family member B; TMEM9 domain family, member B; Tmem9b; Transmembrane protein 9B; UNQ712/PRO1375
Gene Symbols TMEM9B
Host Species Rabbit
Immunogen Recombinant protein of human TMEM9B.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293415, 56674, 56786
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.