missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEPAI Antibody (rPMEPA1/6422), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08259JF646
This item is not returnable.
View return policy
Description
TMEPAI Monoclonal antibody specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TMEPAI | |
| Monoclonal | |
| Janelia Fluor 646 | |
| prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
| Recombinant full-length human TMEPAI protein (Uniprot: Q96W9 ) | |
| 100 μg | |
| Signal Transduction | |
| 56937 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rPMEPA1/6422 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction