missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552598
This item is not returnable.
View return policy
Description
Immunogen sequence: TSTTSDIEVL DHESVISESS ASSRQETTDS KSSLHLMQTS FQLLSASACP EYNRLDDFQK LTESCCSSDA FERIDSFSVQ SLDSRSVSEI NSDDELSGKG YALVPIIVNS STPKSKTVES AEGKSEEVNE TLVIPTEEAE MEESG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
Specifications
| TMF1 | |
| Polyclonal | |
| Unconjugated | |
| TMF1 | |
| 7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114206, 232286, 7110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B9EKI3, P82094 | |
| TMF1 | |
| Recombinant protein corresponding to Human TMF1. Recombinant protein control fragment (Product #RP-89358). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction