missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMIGD2 Antibody (953730), DyLight 755, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB83161IR
This item is not returnable.
View return policy
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TMIGD2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TMIGD2 | |
| Partial recombinant human CD28H protein (amino acids 23-150) [UniProt Q96BF3] | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 953730 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 126259 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction