Learn More
TMIGD2 Antibody (953743), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB83162PECY55
Product Code. 30036285
  • ONLINE EXCLUSIVE PRICE
    € 730.00
    € 689.85 / 0.10mL
    Save € 40.15
    5% Off
Description
TMIGD2 Monoclonal antibody specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMIGD2
Monoclonal
PE-Cyanine5.5
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
126259
Store at 4C in the dark. Do not freeze.
IgG2a
Flow Cytometry
953743
Flow Cytometry
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
Human embryonic kidney cell line HEK-293-derived human TMIGD2/CD28H, Met1-Gly150, Accession # Q96BF3
0.1 mL