missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMIGD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574981
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574981 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574981 Supplier Invitrogen™ Supplier No. PA538055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A recommended positive control is mouse small intestine tissue lysate. At least three isoforms of TMIGD2 are known to exist; this antibody will detect all three. TMIGD2 antibody is not expected to cross-react with TMIGD1.

TMIGD2 (transmembrane and immunoglobulin domain containing 1), also known as immunoglobulin-containing and proline-rich receptor 1 (IGPR1), is novel adhesion molecule that is expressed in multiple tissues, primarily in cells of epithelium and endothelium origins. TMIGD2 is thought to be involved in angiogenesis and regulates cellular morphology, homophilic cell aggregation, and cell-cell interaction. TMIGD2 activity also modulates actin stress fiber formation and focal adhesion and reduces cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMIGD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMIGD2
Gene Accession No. Q96BF3
Gene Alias CD28 homolog; CD28 homologue; CD28H; IGPR1; IGPR-1; Immunoglobulin and proline-rich receptor 1; immunoglobulin-containing and proline-rich receptor 1; immunoglobulin-containing and proline-rich receptor-1; TMIGD2; transmembrane and immunoglobulin domain containing 2; transmembrane and immunoglobulin domain-containing protein 2; transmembrane and immunoglobulin domain-containing protein 2 variant 2; transmembrane and immunoglobulin domain-containing protein 2 variant 3; UNQ3059/PRO9879
Gene Symbols TMIGD2
Host Species Rabbit
Immunogen 18 amino acid peptide near the carboxy terminus of human TMIGD2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126259
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.