Learn More
TMOD1 Mouse anti-Human, Clone: OTI5F8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503174
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TMOD1 gene encodes a member of the tropomodulin family. The encoded protein is an actin-capping protein that regulates tropomyosin by binding to its N-terminus, inhibiting depolymerization and elongation of the pointed end of actin filaments and thereby influencing the structure of the erythrocyte membrane skeleton. Multiple transcript variants encoding the same protein have been found for this gene.Specifications
| TMOD1 | |
| Monoclonal | |
| Unconjugated | |
| TMOD1 | |
| D9S57E, ETMOD, TMOD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7111 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5F8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P28289 | |
| TMOD1 | |
| Full length human recombit protein of human TMOD1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.