missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tmp21/p23 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-57587DL594
This item is not returnable.
View return policy
Description
Tmp21/p23 Polyclonal antibody specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Specifications
| Tmp21/p23 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Primate | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| DyLight 594 | |
| 21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein | |
| A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 50-150). [Swiss-Prot# P49755] | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
| 10972 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction