missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS11D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542875
This item is not returnable.
View return policy
Description
Peptide sequence: RSSFQLLNVE YNSQLNSPAT QEYRTLSGRI ESLITKTFKE SNLRNQFIRA Sequence homology: Guinea Pig: 79%; Horse: 79%; Human: 100%; Rabbit: 79%.
The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
Specifications
| TMPRSS11D | |
| Polyclonal | |
| Unconjugated | |
| TMPRSS11D | |
| Adrenal secretory serine protease; airway spesific trypsin-like protease; airway trypsin like protease; airway trypsin-like protease; AsP; AST; AT; BC020151; HAT; Mat; MGC150587; MGC150588; Rat; TMPRSS11D; Transmembrane protease serine 11D; Transmembrane protease serine 11D catalytic chain; Transmembrane protease serine 11D non-catalytic chain; transmembrane protease, serine 11D; transmembrane serine protease 11D | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60235 | |
| TMPRSS11D | |
| Synthetic peptide directed towards the N-terminal of human TMPRSS11D (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction