missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMPRSS11E Polyclonal Antibody
GREENER_CHOICE

Product Code. 15740102 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15740102 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15740102 Supplier Invitrogen™ Supplier No. PA562306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VLAHMLLICR FHSTEDPETV DKIVQLVLHE KLQDAVGPPK VDPHSVKIKK INKTETDSYL NHCCGTRRSK TLGQSLRIVG GTEVEEGEWP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.

Serine protease which possesses both gelatinolytic and caseinolytic activities. Shows a preference for Arg in the P1 position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS11E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMPRSS11E
Gene Accession No. Q9UL52
Gene Alias DESC1; Serine protease DESC1; TMPRSS11E; TMPRSS11E2; transmembrane protease serine 11E; Transmembrane protease serine 11E catalytic chain; Transmembrane protease serine 11E non-catalytic chain; Transmembrane protease serine 11E2; transmembrane protease, serine 11E; transmembrane protease, serine 11E2; UNQ742/PRO1461
Gene Symbols TMPRSS11E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMPRSS11E. Recombinant protein control fragment (Product #RP-99514).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.