missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMPRSS11F Polyclonal Antibody
GREENER_CHOICE

Product Code. 15753381 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15753381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15753381 Supplier Invitrogen™ Supplier No. PA555461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NKDPTQWIAT FGATITPPAV KRNVRKIILH ENYHRETNEN DIALVQLSTG VEFSNIVQRV CLPDSSIKLP PKTSVFVTGF GSIVDDGPIQ NTLRQARVET ISTDVCNRKD VYDG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.

TMPRSS11F is a type-II transmembrane protease, similar to hepsin (TMPRSS1). TMPRSS11F is a member of a larger family of membrane attached serine proteases, a poorly defined group that includes TMPRSS11A, B, C, D, E, F, Hepsin, Corin, Matriptase-1, 2 and 3. TMPRSS11F has a domain structure of an aminoterminal cytoplasmic domain, followed by a transmembrane domain, a SEA domain (Sea urchin sperm protein, Enterokinase, Agrin), a short spacer, then the trypsin-like serine protease domain. The SEA domain is thought to play a role in carbohydrate binding in the analogous protein sequences where it is found, but its role in TMPRSS11F is unclear. The cleavage of the Arg206-Ile207 bond is thought to liberate the catalytic domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS11F
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMPRSS11F
Gene Accession No. Q6ZWK6
Gene Alias 4631408C20; 4732406D01Rik; airway trypsin-like protease 4; HATL4; TMPRSS11F; Transmembrane protease serine 11F; transmembrane protease, serine 11f
Gene Symbols TMPRSS11F
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMPRSS11F. Recombinant protein control fragment (Product #RP-90611).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389208
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.