missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145223
This item is not returnable.
View return policy
Description
Immunogen sequence: HIHPACLPMH GQTFSLNETC WITGFGKTRE TDDKTSPFLR EVQVNLIDFK KCNDYLVYDS YLTPRMMCAG DL Antigen sequence identity: Mouse 96%; Rat 94%.
This gene encodes a member of the type II transmembrane serine protease family. Transmembrane serine proteases are regulated by protease inhibitors and known to function in development, homeostasis, infection, and tumorigenesis. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| TMPRSS13 | |
| Polyclonal | |
| Unconjugated | |
| TMPRSS13 | |
| membrane-type mosaic serine protease; Mosaic serine protease; MSP; MSPL; MSPS; TMPRSS11; TMPRSS13; Transmembrane protease serine 13; transmembrane protease, serine 11; transmembrane protease, serine 13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYE2 | |
| TMPRSS13 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 429-500. Recombinant protein control fragment (Product #RP-110049). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction