Learn More
Novus Biologicals™ TMPRSS2 Antibody (Regeneron patent anti-TMPRSS2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27959-1mg
 View more versions of this product
Product Code. 30048166
  • € 920.00 / 1mg
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMPRSS2
Monoclonal
Unconjugated
O15393
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Regeneron patent anti-TMPRSS2
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2
TMPRSS2
1 mg
Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites
7113