missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS2 Monoclonal Antibody (CL12511)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546521
This item is not returnable.
View return policy
Description
Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 66%.
TMPRSS2 is a protein that belongs to the serine protease family. The protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. Its gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage.
Specifications
| TMPRSS2 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15393 | |
| TMPRSS2 | |
| Recombinant protein fragment of Human TMPRSS2 (Product #RP-102679) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| CL12511 | |
| Unconjugated | |
| TMPRSS2 | |
| D16Ertd61e; EC 3.4.219; epitheliasin; FLJ41954; MGC6821; Plasmic transmembrane protein X; PP9284; PRSS10; Serine protease 10; Tmprss2; TMPS2; Transmembrane protease serine 2; Transmembrane protease serine 2 catalytic chain; Transmembrane protease serine 2 non-catalytic chain; transmembrane protease, serine 2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction