missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS5 Mouse anti-Human, Clone: OTI4A11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503409
This item is not returnable.
View return policy
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes.Specifications
| TMPRSS5 | |
| Monoclonal | |
| Unconjugated | |
| TMPRSS5 | |
| SPINESIN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80975 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4A11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H3S3 | |
| TMPRSS5 | |
| Full length human recombit protein of human TMPRSS5 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?