missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TMPRSS7 (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
- Sequence: MCHFKLVAIVGYLIRLSIKSIQIEADNCVTDSLTIYDSLLPIRSSILYRICEPTRTLMSFVSTNNLMLVTFKSPHIRRLSGIRAYFEVIPEQKCENTVLVKDITGFEGKISSPYYPSYYPPKCKCTWKFQTSLSTLGIALKFYNYSITKKSMKGCEHGWWEINEHMYCGSYMDHQTIFRVPSPLVHIQLQCSSRLSDKPLLAEYGSYNISQPCPVGSFRCSSGLCVPQAQRCDGVNDCFDESDELFCVSPQPACNTSSFRQHGPLICDGFRDCENGRDEQNCTQSIPCNNRTFKCGNDICFRKQNAKCDGTVDCPDGSDEEGCTCSRSSSALHRIIGGTDTLEGGWPWQVSLHFVGSAYCGASVISREWLLSAAHCFHGNRLSDPTPWTAHLGMYVQGNAKFVSPVRRIVVHEYYNSQTFDYDIALLQLSIAWPETLKQLIQPICIPPTGQRVRSGEKCWVTGWGRRHEADNKGSLVLQQAEVELIDQTLCVSTYGIITSRMLCAGIMSGKRDACKGDSGGPLSCRRKSDGKWILTGIVSWGHGCGRPNFPGVYTRVSNFVPWIHKYVPSLL
Specifications
Specifications
| Accession Number | ADR83226.1 |
| Gene ID (Entrez) | 344805 |
| Name | transmembrane protease, serine 7 |
| Preparation Method | Wheat germ expression system |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Symbol | TMPRSS7 |
| Species | Wheat Germ (in vitro) |
| Buffer | 25 mM Tris-HCl of pH 8.0 containing 2% glycerol |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction