missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMUB1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18424G
This item is not returnable.
View return policy
Description
TMUB1 Polyclonal antibody specifically detects TMUB1 in Human samples. It is validated for Western Blot
Specifications
| TMUB1 | |
| Polyclonal | |
| Western Blot | |
| C7orf21, chromosome 7 open reading frame 21, DULP, Hepatocyte odd protein shuttling protein, HOPS, MGC5442, SB144, transmembrane and ubiquitin-like domain containing 1, transmembrane and ubiquitin-like domain-containing protein 1, Ubiquitin-like protein DULP, Ubiquitin-like protein SB144 | |
| Recognizes region between residues Between Between 50 and 100.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 83590 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction