Learn More
TMUB2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00175PE
This item is not returnable.
View return policy
Description
TMUB2 Polyclonal antibody specifically detects TMUB2 in Human samples. It is validated for ELISA