missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMX4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18462AF647
This item is not returnable.
View return policy
Description
TMX4 Polyclonal antibody specifically detects TMX4 in Human samples. It is validated for Western Blot
Specifications
| TMX4 | |
| Polyclonal | |
| Western Blot | |
| DJ971N18.2, KIAA1162Thioredoxin domain-containing protein 13, PDIA14, protein disulfide isomerase family A, member 14, thioredoxin domain containing 13, thioredoxin-related transmembrane protein 4, TXNDC13 | |
| Recognizes region between residues Between Between 299 and 349.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 56255 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction