missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNC Mouse anti-Human, Clone: T2H5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14284.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human TNC.
- High specificity
- High reactivity
Specifications
| TNC | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| P24821 | |
| TNC | |
| Human breast carcinoma. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| T2H5 | |
| Mouse monoclonal antibody raised against native human TNC. | |
| In 1 mM PBS. | |
| HXB/MGC167029/TN | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3371 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction